Assay Details
| Gene Symbol: Proc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042768.2 | NP_001036233.2 | 1 - 2 | 74 | 70 | |
| GenBank mRNA | BY788141.1 | - | 1 - 2 | 51 | 70 | |
| CA456834.1 | - | 115 | 70 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001042768.2 transcripts.
Set Membership: |
|

Proc
Genomic Map