Assay Details
| Gene Symbol: Iws1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173441.3 | NP_775617.1 | 10 - 11 | 2167 | 99 | |
| GenBank mRNA | AK006925.1 | - | 4 - 5 | 460 | 99 | |
| AK028247.1 | - | 10 - 11 | 2138 | 99 | ||
| AK033013.1 | - | 10 - 11 | 2162 | 99 | ||
| AK136492.1 | - | 6 - 7 | 623 | 99 | ||
| BC068184.1 | - | 9 - 10 | 2753 | 30639737 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iws1
Genomic Map