Assay Details
| Gene Symbol: Iws1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173441.3 | NP_775617.1 | 11 - 12 | 2272 | 59 | |
| GenBank mRNA | AK006925.1 | - | 5 - 6 | 565 | 59 | |
| AK028247.1 | - | 11 - 12 | 2243 | 59 | ||
| AK033013.1 | - | 11 - 12 | 2267 | 59 | ||
| AK136492.1 | - | 7 - 8 | 728 | 59 | ||
| BC068184.1 | - | 10 - 11 | 2858 | 30639737 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iws1
Genomic Map