Assay Details
| Gene Symbol: Iws1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173441.3 | NP_775617.1 | 4 - 5 | 1515 | 86 | |
| GenBank mRNA | AK028247.1 | - | 4 - 5 | 1486 | 86 | |
| AK033013.1 | - | 4 - 5 | 1511 | 86 | ||
| BC068184.1 | - | 4 - 5 | 1455 | 30639737 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iws1
Genomic Map