Assay Details
| Gene Symbol: Iws1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173441.3 | NP_775617.1 | 5 - 6 | 1581 | 93 | |
| GenBank mRNA | AK028247.1 | - | 5 - 6 | 1552 | 93 | |
| AK033013.1 | - | 5 - 6 | 1577 | 93 | ||
| BC068184.1 | - | 5 - 6 | 1521 | 30639737 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Iws1
Genomic Map