Assay Details
Gene Symbol: 1700074P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006951.1 | - | 1 - 2 | 148 | 113 | |
BC061035.1 | - | 1 - 2 | 185 | 6774253 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK006951.1, BC061035.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
