Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 12 - 13 | 1735 | 69 | |
| GenBank mRNA | AB091829.1 | - | 12 - 13 | 1735 | 69 | |
| BC018542.1 | - | 2 - 3 | 238 | 69 | ||
| BC033520.1 | - | 10 - 11 | 1359 | 69 | ||
| BC038009.1 | - | 2 - 3 | 261 | 69 | ||
| BC058953.1 | - | 4 - 5 | 728 | 69 | ||
| BC125377.1 | - | 12 - 13 | 1658 | 40129892 | 69 | |
| BC125379.1 | - | 12 - 13 | 1658 | 40129894 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map