Assay Details
| Gene Symbol: Casd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145398.2 | NP_663373.2 | 16 - 17 | 2133 | 137 | |
| GenBank mRNA | AB091829.1 | - | 16 - 17 | 2133 | 137 | |
| BC018542.1 | - | 6 - 7 | 636 | 137 | ||
| BC033520.1 | - | 14 - 15 | 1757 | 137 | ||
| BC038009.1 | - | 6 - 7 | 659 | 137 | ||
| BC058953.1 | - | 8 - 9 | 1126 | 137 | ||
| BC125377.1 | - | 16 - 17 | 2056 | 40129892 | 137 | |
| BC125379.1 | - | 16 - 17 | 2056 | 40129894 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casd1
Genomic Map