Assay Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC049995.1 | - | 3 - 4 | 497 | 6399201 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgpp2| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC049995.1 | - | 3 - 4 | 497 | 6399201 | 64 |
Set Membership: |
|