Assay Details
| Gene Symbol: Pot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133931.4 | NP_598692.1 | 16 - 17 | 2087 | 123 | |
| GenBank mRNA | AK036052.1 | - | 16 - 17 | 2088 | 123 | |
| AK040864.1 | - | 16 - 17 | 2080 | 123 | ||
| BC016121.1 | - | 16 - 17 | 2049 | 4501733 | 123 | |
More Information
Set Membership: |
|

Pot1a
Genomic Map