Assay Details
| Gene Symbol: Pot1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133931.4 | NP_598692.1 | 2 - 3 | 122 | 88 | |
| GenBank mRNA | AK005318.1 | - | 2 - 3 | 141 | 88 | |
| AK036052.1 | - | 2 - 3 | 123 | 88 | ||
| AK040864.1 | - | 2 - 3 | 115 | 88 | ||
| AK075661.1 | - | 2 - 3 | 113 | 88 | ||
| AK085041.1 | - | 2 - 3 | 113 | 88 | ||
| AK135346.1 | - | 2 - 3 | 92 | 88 | ||
| AK137376.1 | - | 2 - 3 | 92 | 88 | ||
| BC016121.1 | - | 2 - 3 | 84 | 4501733 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK036052.1, AK040864.1, AK085041.1, AK135346.1, AK137376.1, BC016121.1, NM_133931.4 transcripts.
Set Membership: |
|

Pot1a
Genomic Map