Assay Details
| Gene Symbol: Car5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181315.4 | NP_851832.2 | 1 - 2 | 82 | 70 | |
| GenBank mRNA | AF192978.1 | - | 1 - 2 | 103 | 70 | |
| AK046691.1 | - | 1 - 2 | 54 | 70 | ||
| AK085670.1 | - | 1 - 2 | 57 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF192978.1, AK085670.1, NM_181315.4 transcripts.
Set Membership: |
|

Car5b
Genomic Map