Assay Details
| Gene Symbol: Car5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181315.4 | NP_851832.2 | 2 - 3 | 268 | 89 | |
| GenBank mRNA | AF192978.1 | - | 2 - 3 | 289 | 89 | |
| AK085670.1 | - | 2 - 3 | 243 | 89 | ||
| BC034413.1 | - | 2 - 3 | 219 | 4221296 | 89 | |
| BY121187.1 | - | 2 - 3 | 269 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car5b
Genomic Map