Assay Details
| Gene Symbol: Zrsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009453.3 | NP_033479.2 | 10 - 11 | 1287 | 58 | |
| GenBank mRNA | D45205.1 | - | 10 - 11 | 1210 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zrsr2
Genomic Map