Assay Details
| Gene Symbol: Ace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130513.1 | NP_001123985.1 | 11 - 12 | 1774 | 65 | |
| NM_027286.4 | NP_081562.2 | 10 - 11 | 1626 | 65 | ||
| GenBank mRNA | AB053181.1 | - | 10 - 11 | 1630 | 65 | |
| AK135357.1 | - | 5 - 6 | 808 | 65 | ||
| AK141604.1 | - | 10 - 11 | 1475 | 65 | ||
| BC026801.1 | - | 10 - 11 | 1619 | 4236529 | 65 | |
| EF408739.1 | - | 11 - 12 | 1735 | 65 | ||
| EF408740.1 | - | 11 - 12 | 1735 | 65 | ||
| GQ262785.1 | - | 10 - 11 | 1444 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ace2
Genomic Map