Assay Details
| Gene Symbol: Ace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130513.1 | NP_001123985.1 | 10 - 11 | 1629 | 70 | |
| NM_027286.4 | NP_081562.2 | 9 - 10 | 1481 | 70 | ||
| GenBank mRNA | AB053181.1 | - | 9 - 10 | 1485 | 70 | |
| AK135357.1 | - | 4 - 5 | 663 | 70 | ||
| AK141604.1 | - | 9 - 10 | 1330 | 70 | ||
| BC026801.1 | - | 9 - 10 | 1474 | 4236529 | 70 | |
| EF408739.1 | - | 10 - 11 | 1590 | 70 | ||
| EF408740.1 | - | 10 - 11 | 1590 | 70 | ||
| GQ262785.1 | - | 9 - 10 | 1299 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ace2
Genomic Map