Assay Details
| Gene Symbol: Pax4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011038.2 | NP_035168.1 | 9 - 10 | 1036 | 80 | |
| GenBank mRNA | AB008912.1 | - | 9 - 10 | 1011 | 80 | |
| AB010557.1 | - | 9 - 10 | 979 | 80 | ||
| AB010558.1 | - | 9 - 10 | 994 | 80 | ||
| AF031150.1 | - | 9 - 10 | 1069 | 80 | ||
| AF104231.1 | - | 8 - 9 | 791 | 80 | ||
| AF145233.1 | - | 9 - 10 | 983 | 80 | ||
| BC131906.1 | - | 9 - 10 | 947 | 40130183 | 80 | |
| BC131908.1 | - | 9 - 10 | 945 | 40130185 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax4
Genomic Map