| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
TC0600001967.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_001159925.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_001159925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_001159926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_001159926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_011038.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015429.mm.1 |
NM_011038.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
TC0600001967.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_001159925.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_001159925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_001159926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_001159926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_011038.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015428.mm.1 |
NM_011038.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
TC0600001967.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_001159925.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_001159925.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_001159926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_001159926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_011038.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600015431.mm.1 |
NM_011038.2 |
Refseq |