Assay Details
| Gene Symbol: Pax4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159925.1 | NP_001153397.1 | 2 - 3 | 278 | 81 | |
| NM_001159926.1 | NP_001153398.1 | 2 - 3 | 278 | 81 | ||
| NM_011038.2 | NP_035168.1 | 2 - 3 | 278 | 81 | ||
| GenBank mRNA | AB008912.1 | - | 2 - 3 | 253 | 81 | |
| AB010557.1 | - | 2 - 3 | 221 | 81 | ||
| AB010558.1 | - | 2 - 3 | 236 | 81 | ||
| AF031150.1 | - | 2 - 3 | 311 | 81 | ||
| AF145233.1 | - | 2 - 3 | 225 | 81 | ||
| BC131906.1 | - | 2 - 3 | 189 | 40130183 | 81 | |
| BC131908.1 | - | 2 - 3 | 187 | 40130185 | 81 | |
| BC145168.1 | - | 2 - 3 | 189 | 9053719 | 81 | |
| BC145169.1 | - | 2 - 3 | 189 | 9053720 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB008912.1, AB010557.1, AB010558.1, AF031150.1, AF145233.1, BC131906.1, BC131908.1, BC145168.1, BC145169.1, NM_001159925.1, NM_001159926.1, NM_011038.2 transcripts.
Set Membership: |
|

Pax4
Genomic Map