Assay Details
| Gene Symbol: Pax4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159925.1 | NP_001153397.1 | 3 - 4 | 404 | 62 | |
| NM_001159926.1 | NP_001153398.1 | 3 - 4 | 404 | 62 | ||
| NM_011038.2 | NP_035168.1 | 3 - 4 | 404 | 62 | ||
| GenBank mRNA | AB008912.1 | - | 3 - 4 | 379 | 62 | |
| AB010557.1 | - | 3 - 4 | 347 | 62 | ||
| AB010558.1 | - | 3 - 4 | 362 | 62 | ||
| AF031150.1 | - | 3 - 4 | 437 | 62 | ||
| AF104231.1 | - | 2 - 3 | 159 | 62 | ||
| AF145233.1 | - | 3 - 4 | 351 | 62 | ||
| BC131906.1 | - | 3 - 4 | 315 | 40130183 | 62 | |
| BC131908.1 | - | 3 - 4 | 313 | 40130185 | 62 | |
| BC145168.1 | - | 3 - 4 | 315 | 9053719 | 62 | |
| BC145169.1 | - | 3 - 4 | 315 | 9053720 | 62 | |
| Y09584.1 | - | 1 - 2 | 43 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pax4
Genomic Map