Assay Details
| Gene Symbol: Pax4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159925.1 | NP_001153397.1 | 6 - 7 | 830 | 76 | |
| NM_001159926.1 | NP_001153398.1 | 6 - 7 | 830 | 76 | ||
| NM_011038.2 | NP_035168.1 | 6 - 7 | 830 | 76 | ||
| GenBank mRNA | AB008912.1 | - | 6 - 7 | 805 | 76 | |
| AB010557.1 | - | 6 - 7 | 773 | 76 | ||
| AB010558.1 | - | 6 - 7 | 788 | 76 | ||
| AF031150.1 | - | 6 - 7 | 863 | 76 | ||
| AF104231.1 | - | 5 - 6 | 585 | 76 | ||
| AF145233.1 | - | 6 - 7 | 777 | 76 | ||
| BC131906.1 | - | 6 - 7 | 741 | 40130183 | 76 | |
| BC131908.1 | - | 6 - 7 | 739 | 40130185 | 76 | |
| BC145168.1 | - | 6 - 7 | 741 | 9053719 | 76 | |
| BC145169.1 | - | 6 - 7 | 741 | 9053720 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pax4
Genomic Map