Assay Details
| Gene Symbol: Magi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170745.1 | NP_001164216.1 | 20 - 21 | 3806 | 96 | |
| NM_001170746.1 | NP_001164217.1 | 22 - 23 | 4013 | 96 | ||
| NM_015823.3 | NP_056638.1 | 21 - 22 | 3848 | 96 | ||
| GenBank mRNA | AK122349.1 | - | 20 - 21 | 3308 | 96 | |
| AK147530.1 | - | 20 - 21 | 3807 | 96 | ||
| BC059005.1 | - | 21 - 22 | 3505 | 6847238 | 96 | |
More Information
Set Membership: |
|

Magi2
Genomic Map