Assay Details
| Gene Symbol: Magi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170745.1 | NP_001164216.1 | 7 - 8 | 1304 | 73 | |
| NM_001170746.1 | NP_001164217.1 | 8 - 9 | 1469 | 73 | ||
| NM_015823.3 | NP_056638.1 | 7 - 8 | 1304 | 73 | ||
| GenBank mRNA | AK039336.1 | - | 4 - 5 | 289 | 73 | |
| AK122349.1 | - | 6 - 7 | 764 | 73 | ||
| AK147530.1 | - | 7 - 8 | 1305 | 73 | ||
| BC059005.1 | - | 7 - 8 | 961 | 6847238 | 73 | |
More Information
Set Membership: |
|

Magi2
Genomic Map