Assay Details
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC055928.1 | - | 1 - 2 | 90 | 3995231 | 94 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055928.1 transcripts.
Set Membership: |
|

Ino80e
Genomic Map