Assay Details
| Gene Symbol: Arhgap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285785.1 | NP_001272714.1 | 8 - 9 | 1200 | 94 | |
| NM_146011.2 | NP_666123.2 | 7 - 8 | 639 | 94 | ||
| GenBank mRNA | AK047280.1 | - | 7 - 8 | 584 | 94 | |
| AK049245.1 | - | 4 - 5 | 420 | 94 | ||
| AK083474.1 | - | 4 - 5 | 549 | 94 | ||
| AK089051.1 | - | 7 - 8 | 591 | 94 | ||
| AK167738.1 | - | 8 - 9 | 777 | 94 | ||
| BC024535.1 | - | 7 - 8 | 634 | 4987970 | 94 | |
| BC027374.1 | - | 7 - 8 | 610 | 4947306 | 94 | |
| DQ498128.1 | - | 8 - 9 | 1048 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047280.1 transcripts.
Set Membership: |
|

Arhgap9
Genomic Map