Assay Details
| Gene Symbol: Stac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177707.3 | NP_808375.1 | 11 - 12 | 1347 | 67 | |
| GenBank mRNA | AK036516.1 | - | 11 - 12 | 1348 | 67 | |
| BC067208.1 | - | 11 - 12 | 1127 | 30286602 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536265 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536268 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618310 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620178 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | AK038830.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | AK048558.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | AK090183.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | AK141582.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | BC027575.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378453 | NM_153597.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 564 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119331 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536207 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27536178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07230457 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119331 | - | - |
| Whole Mouse Genome G4122A | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177261 | NM_177707.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177261 | AK036516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P177261 | BC067208.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115558 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119331 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08730346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08730345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08575126 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08736910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08575122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08736911 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536264 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536265 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536268 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27536263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08487366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08730345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08575125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08783373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08245878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08260658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08339501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08487364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08622966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536209 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10366944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366941 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458087_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10366938 | - | - |
| Mouse Genome 430 2.0 Array | 1458087_at | - | - |
| Mouse Genome 430 2.0 Array | 1458087_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237611_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237611_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011222.mm.1 | NM_177707.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011222.mm.1 | NM_177707.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011222.mm.1 | TC1000001565.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1458087_PM_at | - | - |
| Mouse Expression Set 430 | 1458087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458087_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458087_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 462519 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1500465 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500465 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724841 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724841 | - | - |
| mouse-6 v1.1 (Illumina) | scl38320.11.1_107-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500465 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38320.11.1_107-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500465 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2724841 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2724841 | - | - |

Stac3
Genomic Map