| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600017314.mm.1 |
TC0600002200.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017314.mm.1 |
NM_001146689.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017314.mm.1 |
NM_001146689.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017314.mm.1 |
NM_007971.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017314.mm.1 |
NM_007971.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017315.mm.1 |
TC0600002200.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017315.mm.1 |
NM_001146689.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017315.mm.1 |
NM_001146689.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017315.mm.1 |
NM_007971.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017315.mm.1 |
NM_007971.2 |
Refseq |