Assay Details
| Gene Symbol: Ddah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016765.3 | NP_058045.1 | 6 - 7 | 1020 | 102 | |
| GenBank mRNA | AF004106.1 | - | 6 - 7 | 954 | 102 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ddah2
Genomic Map