Assay Details
| Gene Symbol: Ltb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008518.2 | NP_032544.1 | 2 - 3 | 500 | 121 | |
| GenBank mRNA | BC114919.1 | - | 2 - 3 | 480 | 121 | |
| U16985.1 | - | 2 - 3 | 485 | 121 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158191 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158193 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41793 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | U16985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | U16985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158195 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31158186 | - | - |
| Whole Mouse Genome G4122A | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | U16985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | BC064062.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | BC114919.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302358 | NM_008518.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140577 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13231047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13231062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158192 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158190 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158193 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158188 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31158191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31158166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13231054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13516002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13199204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13164356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13231065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13231061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31158190 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444755 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419135_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444752 | - | - |
| Mouse Genome 430 2.0 Array | 1419135_at | - | - |
| Mouse Genome 430 2.0 Array | 1419135_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16994_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16994_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006869.mm.1 | NM_008518.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006869.mm.1 | NM_008518.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006869.mm.1 | TC1700000673.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006868.mm.1 | NM_008518.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006868.mm.1 | NM_008518.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006868.mm.1 | TC1700000673.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419135_at | - | - |
| Mouse Expression Set 430 | 1419135_PM_at | - | - |
| Murine Genome U74v2 Set | 102940_PM_at | - | - |
| Murine Genome U74v2 Set | 102940_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419135_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419135_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419135_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419135_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 450136 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3990672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003990672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258283 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258283 | - | - |
| mouse-6 v1.1 (Illumina) | scl50784.3.81_39-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3990672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50784.3.81_39-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3990672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1258283 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258283 | - | - |

Ltb
Genomic Map