Assay Details
| Gene Symbol: Copb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033370.3 | NP_203534.1 | 1 - 2 | 75 | 132 | |
| GenBank mRNA | AK011453.1 | - | 1 - 2 | 101 | 132 | |
| AK082977.1 | - | 1 - 2 | 89 | 132 | ||
| AK135168.1 | - | 1 - 2 | 108 | 132 | ||
| AK149828.1 | - | 1 - 2 | 93 | 132 | ||
| AK152222.1 | - | 1 - 2 | 102 | 132 | ||
| AK161544.1 | - | 1 - 2 | 102 | 132 | ||
| AK172214.1 | - | 1 - 2 | 77 | 132 | ||
| BC030837.1 | - | 1 - 2 | 75 | 4022216 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149828.1, AK152222.1, AK161544.1, AK172214.1, BC030837.1, NM_033370.3 transcripts.
Set Membership: |
|

Copb1
Genomic Map