Assay Details
| Gene Symbol: Olfr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146370.1 | NP_666482.1 | 1 | 262 | 116 | |
| GenBank mRNA | AF102521.1 | - | 1 | 76 | 116 | |
| BC120658.1 | - | 1 | 294 | 40129581 | 116 | |
| BC120660.1 | - | 1 | 294 | 40129583 | 116 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr47
Genomic Map