Assay Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016756.4 | NP_058036.1 | 3 - 4 | 534 | 57 | |
| NM_183417.3 | NP_904326.1 | 3 - 4 | 534 | 57 | ||
| GenBank mRNA | AJ223732.1 | - | 3 - 4 | 436 | 57 | |
| AK042462.1 | - | 2 - 3 | 1195 | 57 | ||
| AK146845.1 | - | 3 - 4 | 514 | 57 | ||
| AK148017.1 | - | 3 - 4 | 473 | 57 | ||
| AK151979.1 | - | 3 - 4 | 484 | 57 | ||
| AK152922.1 | - | 3 - 4 | 487 | 57 | ||
| AK153983.1 | - | 3 - 4 | 486 | 57 | ||
| AK154997.1 | - | 3 - 4 | 486 | 57 | ||
| AK179714.1 | - | 2 - 3 | 206 | 57 | ||
| AK184593.1 | - | 2 - 3 | 203 | 57 | ||
| AK186779.1 | - | 2 - 3 | 205 | 57 | ||
| AK187629.1 | - | 3 - 4 | 270 | 57 | ||
| AK189612.1 | - | 3 - 4 | 270 | 57 | ||
| AK210959.1 | - | 2 - 3 | 205 | 57 | ||
| AK219393.1 | - | 2 - 3 | 205 | 57 | ||
| BC005654.1 | - | 3 - 4 | 495 | 3709730 | 57 | |
| BC026145.1 | - | 2 - 3 | 1240 | 57 | ||
| U63337.1 | - | 3 - 4 | 462 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk2
Genomic Map