Assay Details
| Gene Symbol: Cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016756.4 | NP_058036.1 | 5 - 6 | 812 | 67 | |
| GenBank mRNA | AK152922.1 | - | 5 - 6 | 765 | 67 | |
| BC026145.1 | - | 4 - 5 | 1518 | 67 | ||
| U63337.1 | - | 5 - 6 | 740 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk2
Genomic Map