Assay Details
| Gene Symbol: Nabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027257.1 | NP_081533.1 | 6 - 7 | 525 | 82 | |
| GenBank mRNA | AK011703.1 | - | 5 - 6 | 493 | 82 | |
| BC026942.1 | - | 6 - 7 | 525 | 5011571 | 82 | |
| DQ158906.1 | - | 6 - 7 | 525 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541275 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541274 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004588 | NM_027257.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004588 | BC026942.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004588 | DQ158906.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67748 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150273 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004588 | NM_027257.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004588 | BC026942.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004588 | DQ158906.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541243 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27541257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02247291 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07233038 | - | - |
| Whole Mouse Genome G4122A | A_51_P294883 | NM_027257.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294883 | AK011703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294883 | BC026942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P294883 | DQ158906.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121185 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02247291 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08358398 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08358400 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08812405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08562967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08497089 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08497098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08812419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08497111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08358397 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541274 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27541275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27541301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08497093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08812394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08969478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08497101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08497103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27541267 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373394 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373391 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451291_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373388 | - | - |
| Mouse Genome 430 2.0 Array | 1451291_at | - | - |
| Mouse Genome 430 2.0 Array | 1451291_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69917_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69917_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000023560.mm.1 | NM_027257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023560.mm.1 | NM_027257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023560.mm.1 | TC1000003144.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023559.mm.1 | NM_027257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023559.mm.1 | NM_027257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023559.mm.1 | TC1000003144.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000023561.mm.1 | NM_027257.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023561.mm.1 | NM_027257.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000023561.mm.1 | TC1000003144.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1451291_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451291_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1940341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1940341 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940341 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2593852 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2593852 | - | - |
| mouse-6 v1.1 (Illumina) | scl37365.9.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940341 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37365.9.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940341 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2593852 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2837156 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837156 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2593852 | - | - |

Nabp2
Genomic Map