Assay Details
| Gene Symbol: Rab5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177411.4 | NP_803130.1 | 5 - 6 | 711 | 117 | |
| GenBank mRNA | AB232594.1 | - | 4 - 5 | 572 | 117 | |
| AK081251.1 | - | 5 - 6 | 707 | 117 | ||
| BC137574.1 | - | 4 - 5 | 623 | 8860593 | 117 | |
| BC137575.1 | - | 4 - 5 | 623 | 8860594 | 117 | |
| X84239.1 | - | 4 - 5 | 580 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab5b
Genomic Map