Assay Details
| Gene Symbol: Ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024180.5 | NP_077142.1 | 1 - 2 | 92 | 59 | |
| GenBank mRNA | AK002583.1 | - | 1 - 2 | 55 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002583.1, NM_024180.5 transcripts.
Set Membership: |
|

Ormdl2
Genomic Map