Assay Details
| Gene Symbol: Emc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025736.2 | NP_080012.1 | 5 - 6 | 427 | 118 | |
| GenBank mRNA | AF332065.1 | - | 5 - 6 | 355 | 118 | |
| AF332066.1 | - | 5 - 6 | 355 | 118 | ||
| AK014989.1 | - | 5 - 6 | 428 | 118 | ||
| AK015968.1 | - | 5 - 6 | 393 | 118 | ||
| AK172887.1 | - | 5 - 6 | 372 | 118 | ||
| BC004716.1 | - | 5 - 6 | 385 | 3582196 | 118 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Emc2
Genomic Map