Assay Details
| Gene Symbol: Trhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013696.2 | NP_038724.1 | 1 - 2 | 250 | 115 | |
| GenBank mRNA | AK134591.1 | - | 1 - 2 | 251 | 115 | |
| BB653069.1 | - | 1 - 2 | 251 | 115 | ||
| BC109005.1 | - | 1 - 2 | 135 | 40048176 | 115 | |
| BC109006.1 | - | 1 - 2 | 135 | 40048178 | 115 | |
| BC128269.1 | - | 1 - 2 | 135 | 40048433 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134591.1, BC109005.1, BC109006.1, BC128269.1, NM_013696.2 transcripts.
Set Membership: |
|

Trhr
Genomic Map