Assay Details
| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284519.1 | NP_001271448.1 | 2 - 3 | 248 | 98 | |
| GenBank mRNA | AB061277.1 | - | 2 - 3 | 232 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB061277.1, NM_001284519.1 transcripts.
Set Membership: |
|

Cmah
Genomic Map