Assay Details
| Gene Symbol: Casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042072.1 | - | 1 - 2 | 92 | 59 | |
| AK133829.1 | - | 1 - 2 | 101 | 59 | ||
| BC034262.1 | - | 1 - 2 | 158 | 4460089 | 59 | |
| BY074786.1 | - | 1 - 2 | 214 | 59 | ||
| D28492.1 | - | 1 - 2 | 81 | 59 | ||
| Y13085.1 | - | 1 - 2 | 160 | 59 | ||
More Information
Set Membership: |
|

Casp2
Genomic Map