Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013160.mm.1 |
NR_045436.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013160.mm.1 |
NR_045436.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013160.mm.1 |
NR_045437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013160.mm.1 |
NR_045437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013160.mm.1 |
TC1700001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013161.mm.1 |
NR_045436.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013161.mm.1 |
NR_045436.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013161.mm.1 |
NR_045437.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013161.mm.1 |
NR_045437.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013161.mm.1 |
TC1700001473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013158.mm.1 |
NR_045436.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013158.mm.1 |
NR_045436.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013158.mm.1 |
TC1700001473.mm.1 |
MTA TC |