Assay Details
| Gene Symbol: Tac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199971.1 | NP_001186900.1 | 6 - 7 | 559 | 149 | |
| NM_009312.2 | NP_033338.2 | 5 - 6 | 387 | 149 | ||
| GenBank mRNA | AF186116.1 | - | 5 - 6 | 353 | 149 | |
| BC031348.1 | - | 5 - 6 | 384 | 149 | ||
| BC051476.1 | - | 5 - 6 | 393 | 1533442 | 149 | |
| BQ175657.1 | - | 2 - 3 | 149 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tac2
Genomic Map