Assay Details
| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008921.2 | NP_032947.1 | 1 - 2 | 135 | 120 | |
| GenBank mRNA | AK088557.1 | - | 1 - 2 | 155 | 120 | |
| AK089063.1 | - | 1 - 2 | 160 | 120 | ||
| AK133878.1 | - | 1 - 2 | 132 | 120 | ||
| AK160831.1 | - | 1 - 2 | 155 | 120 | ||
| AK169632.1 | - | 1 - 2 | 132 | 120 | ||
| BC089544.1 | - | 1 - 2 | 174 | 30504228 | 120 | |
| D13544.1 | - | 1 - 2 | 135 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim1
Genomic Map