Assay Details
| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008921.2 | NP_032947.1 | 5 - 6 | 606 | 143 | |
| GenBank mRNA | AK011456.1 | - | 5 - 6 | 588 | 143 | |
| AK088557.1 | - | 5 - 6 | 626 | 143 | ||
| AK089063.1 | - | 5 - 6 | 631 | 143 | ||
| AK133878.1 | - | 5 - 6 | 603 | 143 | ||
| AK169632.1 | - | 5 - 6 | 603 | 143 | ||
| BC089544.1 | - | 5 - 6 | 645 | 30504228 | 143 | |
| D13544.1 | - | 5 - 6 | 606 | 143 | ||
| J04620.1 | - | 5 - 6 | 665 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim1
Genomic Map