Assay Details
| Gene Symbol: Prim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008921.2 | NP_032947.1 | 8 - 9 | 872 | 62 | |
| GenBank mRNA | AK011456.1 | - | 8 - 9 | 854 | 62 | |
| AK088557.1 | - | 8 - 9 | 892 | 62 | ||
| AK089063.1 | - | 8 - 9 | 897 | 62 | ||
| AK133878.1 | - | 8 - 9 | 869 | 62 | ||
| AK160831.1 | - | 6 - 7 | 2634 | 62 | ||
| AK169632.1 | - | 8 - 9 | 869 | 62 | ||
| BC089544.1 | - | 8 - 9 | 911 | 30504228 | 62 | |
| D13544.1 | - | 8 - 9 | 872 | 62 | ||
| J04620.1 | - | 8 - 9 | 931 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prim1
Genomic Map