Assay Details
| Gene Symbol: Ndufa4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098789.1 | NP_001092259.1 | 1 - 2 | 101 | 66 | |
| GenBank mRNA | AK131609.1 | - | 1 - 2 | 73 | 66 | |
| AK131662.1 | - | 1 - 2 | 70 | 66 | ||
| BC064011.1 | - | 1 - 2 | 62 | 66 | ||
| BC099496.1 | - | 1 - 2 | 62 | 30847512 | 66 | |
| BY336656.1 | - | 1 - 2 | 103 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971244 | BC064011.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971244 | BC099496.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971244 | NM_001098789.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971244 | BC064011.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971244 | BC099496.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971244 | NM_001098789.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136249 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394778 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394787 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394821 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08394786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27536392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08394821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536314 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08394820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08622969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08394787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08622957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27536268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08243101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10366955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10366954 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434905_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10366951 | - | - |
| Mouse Genome 430 2.0 Array | 1434905_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434905_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407790_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 407790_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000011226.mm.1 | NM_001098789.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011226.mm.1 | NM_001098789.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011226.mm.1 | TC1000001566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011225.mm.1 | NM_001098789.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011225.mm.1 | NM_001098789.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011225.mm.1 | TC1000001566.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000011227.mm.1 | NM_001098789.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011227.mm.1 | NM_001098789.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000011227.mm.1 | TC1000001566.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1434905_PM_at | - | - |
| Mouse Expression Set 430 | 1434905_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434905_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434905_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 852251 | - | - |

Ndufa4l2
Genomic Map