Assay Details
| Gene Symbol: Tmem194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113211.1 | NP_001106682.1 | 5 - 6 | 664 | 112 | |
| NM_173732.3 | NP_776093.1 | 5 - 6 | 664 | 112 | ||
| GenBank mRNA | AK090031.1 | - | 5 - 6 | 643 | 112 | |
| AK129110.1 | - | 5 - 6 | 661 | 112 | ||
| AK132974.1 | - | 1133 | 112 | |||
| BC030440.1 | - | 5 - 6 | 675 | 5373527 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem194
Genomic Map