| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000011057.mm.1 |
NM_001113211.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011057.mm.1 |
NM_001113211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011057.mm.1 |
TC1000001554.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011059.mm.1 |
NM_001113211.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011059.mm.1 |
NM_001113211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011059.mm.1 |
TC1000001554.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011058.mm.1 |
NM_001113211.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011058.mm.1 |
NM_001113211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011058.mm.1 |
TC1000001554.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011056.mm.1 |
NM_001113211.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011056.mm.1 |
NM_001113211.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011056.mm.1 |
TC1000001554.mm.1 |
MTA TC |