Assay Details
| Gene Symbol: Stat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009284.2 | NP_033310.2 | 18 - 19 | 2344 | 70 | |
| GenBank mRNA | AF481808.1 | - | 17 - 18 | 2059 | 70 | |
| AF481809.1 | - | 17 - 18 | 2059 | 70 | ||
| AK135911.1 | - | 1 - 2 | 106 | 70 | ||
| AK149563.1 | - | 18 - 19 | 2305 | 70 | ||
| AK163053.1 | - | 18 - 19 | 2345 | 70 | ||
| BC029318.1 | - | 5 - 6 | 533 | 70 | ||
| BC127054.1 | - | 18 - 19 | 2134 | 40109712 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stat6
Genomic Map