Assay Details
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194262.2 | NP_919238.1 | 12 - 13 | 1265 | 74 | |
| NM_198122.2 | NP_937755.1 | 12 - 13 | 1265 | 74 | ||
| GenBank mRNA | AK031062.1 | - | 12 - 13 | 1248 | 74 | |
| AK041414.1 | - | 12 - 13 | 1262 | 74 | ||
| AK080355.1 | - | 2 - 3 | 125 | 74 | ||
| AK136105.1 | - | 12 - 13 | 1149 | 74 | ||
| AK161669.1 | - | 12 - 13 | 1251 | 74 | ||
| BC137783.1 | - | 11 - 12 | 1027 | 8860802 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arid4b
Genomic Map